Learn More
ABclonal Technology MATN2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A163971000UL
Encompass_Preferred
This gene encodes a member of the von Willebrand factor A domain containing protein family. This family of proteins is thought to be involved in the formation of filamentous networks in the extracellular matrices of various tissues. This protein contains five von Willebrand factor A domains. The specific function of this gene has not yet been determined. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-09700