Learn More
ABclonal Technology OCM2 Rabbit pAb
SDP
Catalog No. 5000110765
Encompass_Preferred
This gene is similar to the oncomodulin gene, a high-affinity calcium ion-binding protein that belongs to the superfamily of calmodulin proteins, also known as the EF-hand proteins.
Catalog No. 50-001-10765 Supplier ABclonal Technology Supplier No. A16410500UL
This gene is similar to the oncomodulin gene, a high-affinity calcium ion-binding protein that belongs to the superfamily of calmodulin proteins, also known as the EF-hand proteins.