Learn More
ABclonal Technology RANBP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A164221000UL
Encompass_Preferred
This gene encodes a protein that forms a complex with Ras-related nuclear protein (Ran) and metabolizes guanoside triphosphate (GTP). This complex participates in the regulation of the cell cycle by controlling transport of proteins and nucleic acids into the nucleus. There are multiple pseudogenes for this gene on chromosomes 9, 12, 17, and X. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-11955