Learn More
ABclonal Technology INPP4B Rabbit pAb
SDP
Supplier:  ABclonal Technology A164611000UL
Encompass_Preferred
INPP4B encodes the inositol polyphosphate 4-phosphatase type II, one of the enzymes involved in phosphatidylinositol signaling pathways. This enzyme removes the phosphate group at position 4 of the inositol ring from inositol 3,4-bisphosphate. There is limited data to suggest that the human type II enzyme is subject to alternative splicing, as has been established for the type I enzyme.
Catalog No. 50-001-10760