Learn More
ABclonal Technology Fetuin A/AHSG Rabbit pAb
SDP
Supplier:  ABclonal Technology A1647500UL
Encompass_Preferred
The protein encoded by this gene is a negatively-charged serum glycoprotein that is synthesized by hepatocytes. The encoded protein consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several processes, including endocytosis, brain development, and the formation of bone tissue. Defects in this gene are a cause of susceptibility to leanness.
Catalog No. 50-001-11267