Learn More
ABclonal Technology BCLAF1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A164731000UL
Encompass_Preferred
This gene encodes a transcriptional repressor that interacts with several members of the BCL2 family of proteins. Overexpression of this protein induces apoptosis, which can be suppressed by co-expression of BCL2 proteins. The protein localizes to dot-like structures throughout the nucleus, and redistributes to a zone near the nuclear envelope in cells undergoing apoptosis. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-10742