Learn More
ABclonal Technology GPR82 Rabbit pAb
SDP
Supplier:  ABclonal Technology A165161000UL
Encompass_Preferred
The protein encoded by this gene is an orphan G protein-coupled receptor of unknown function. The encoded protein is a member of a family of proteins that contain seven transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
Catalog No. 50-001-13453