Learn More
ABclonal Technology ITSN2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A165251000UL
Encompass_Preferred
This gene encodes a cytoplasmic protein which contains SH3 domains. This protein is a member of a family of proteins involved in clathrin-mediated endocytosis. Intersectin 2 is thought to regulate the formation of clathrin-coated vesicles and also may function in the induction of T cell antigen receptor (TCR) endocytosis.
Catalog No. 50-001-13320