Learn More
ABclonal Technology POP5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A165301000UL
Encompass_Preferred
Enables ribonuclease P RNA binding activity. Contributes to ribonuclease P activity. Involved in tRNA 5-leader removal. Located in nucleolus. Part of multimeric ribonuclease P complex and ribonuclease MRP complex.
Catalog No. 50-001-13621