Learn More
ABclonal Technology PCF11 Rabbit pAb
SDP
Catalog No. 5000111463
Encompass_Preferred
The protein encoded by this gene binds to CLP1 to form pre-mRNA cleavage factor IIm. The encoded protein is necessary for efficient Pol II transcription termination and may be involved in degradation of the 3 product of polyA site cleavage.
Catalog No. 50-001-11463 Supplier ABclonal Technology Supplier No. A16533500UL
The protein encoded by this gene binds to CLP1 to form pre-mRNA cleavage factor IIm. The encoded protein is necessary for efficient Pol II transcription termination and may be involved in degradation of the 3 product of polyA site cleavage.