Learn More
ABclonal Technology SOX18 Rabbit pAb
SDP
Supplier:  ABclonal Technology A165361000UL
Encompass_Preferred
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. This protein plays a role in hair, blood vessel, and lymphatic vessel development. Mutations in this gene have been associated with recessive and dominant forms of hypotrichosis-lymphedema-telangiectasia.
Catalog No. 50-001-12863