Learn More
ABclonal Technology CHDH Rabbit pAb
SDP
Catalog No. 5000113284
Encompass_Preferred
The protein encoded by this gene is a choline dehydrogenase that localizes to the mitochondrion. Variations in this gene can affect susceptibility to choline deficiency. A few transcript variants have been found for this gene, but the full-length nature of only one has been characterized to date.
Catalog No. 50-001-13284 Supplier ABclonal Technology Supplier No. A16545500UL
The protein encoded by this gene is a choline dehydrogenase that localizes to the mitochondrion. Variations in this gene can affect susceptibility to choline deficiency. A few transcript variants have been found for this gene, but the full-length nature of only one has been characterized to date.