Learn More
ABclonal Technology ACBD3 Rabbit pAb
SDP
Catalog No. 5000113309
Encompass_Preferred
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is involved in the maintenance of Golgi structure and function through its interaction with the integral membrane protein giantin. It may also be involved in the hormonal regulation of steroid formation.
Catalog No. 50-001-13309 Supplier ABclonal Technology Supplier No. A16568500UL
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is involved in the maintenance of Golgi structure and function through its interaction with the integral membrane protein giantin. It may also be involved in the hormonal regulation of steroid formation.