Learn More
ABclonal Technology Steroidogenic factor-1 (SF-1/NR5A1) Rabbit pAb
SDP
Catalog No. 5000109433
Encompass_Preferred
The protein encoded by this gene is a transcriptional activator involved in sex determination. The encoded protein binds DNA as a monomer. Defects in this gene are a cause of XY sex reversal with or without adrenal failure as well as adrenocortical insufficiency without ovarian defect.
Catalog No. 50-001-09433 Supplier ABclonal Technology Supplier No. A16571000UL
The protein encoded by this gene is a transcriptional activator involved in sex determination. The encoded protein binds DNA as a monomer. Defects in this gene are a cause of XY sex reversal with or without adrenal failure as well as adrenocortical insufficiency without ovarian defect.