Learn More
ABclonal Technology STARD3NL Rabbit pAb
SDP
Catalog No. 5000109477
Encompass_Preferred
This gene encodes a late-endosomal protein that contains a conserved MENTAL (MLN64 N-terminal) domain. The encoded protein binds cholesterol molecules and may play a role in endosomal cholesterol transport through interactions with metastatic lymph node protein 64 (MLN64).
Catalog No. 50-001-09477 Supplier ABclonal Technology Supplier No. A16579500UL
This gene encodes a late-endosomal protein that contains a conserved MENTAL (MLN64 N-terminal) domain. The encoded protein binds cholesterol molecules and may play a role in endosomal cholesterol transport through interactions with metastatic lymph node protein 64 (MLN64).