Learn More
ABclonal Technology SPIRE2 Rabbit pAb
SDP
Catalog No. 5000110579
Encompass_Preferred
Predicted to enable actin binding activity. Involved in establishment of meiotic spindle localization, formin-nucleated actin cable assembly, and positive regulation of double-strand break repair. Predicted to be located in cytoskeleton, cytosol, and plasma membrane. Predicted to be active in cell cortex and cytoplasmic vesicle membrane. Predicted to colocalize with cleavage furrow.
Catalog No. 50-001-10579 Supplier ABclonal Technology Supplier No. A165821000UL
Predicted to enable actin binding activity. Involved in establishment of meiotic spindle localization, formin-nucleated actin cable assembly, and positive regulation of double-strand break repair. Predicted to be located in cytoskeleton, cytosol, and plasma membrane. Predicted to be active in cell cortex and cytoplasmic vesicle membrane. Predicted to colocalize with cleavage furrow.