Learn More
ABclonal Technology SPIRE2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16582500UL
Encompass_Preferred
Predicted to enable actin binding activity. Involved in establishment of meiotic spindle localization, formin-nucleated actin cable assembly, and positive regulation of double-strand break repair. Predicted to be located in cytoskeleton, cytosol, and plasma membrane. Predicted to be active in cell cortex and cytoplasmic vesicle membrane. Predicted to colocalize with cleavage furrow.
Catalog No. 50-001-09432