Learn More
ABclonal Technology ZGPAT Rabbit pAb
Supplier: ABclonal Technology A165831000UL
Enables DNA-binding transcription repressor activity, RNA polymerase II-specific and RNA polymerase II cis-regulatory region sequence-specific DNA binding activity. Involved in negative regulation of epidermal growth factor-activated receptor activity and negative regulation of transcription by RNA polymerase II. Located in nucleoplasm and plasma membrane.