Learn More
ABclonal Technology NRN1L Rabbit pAb
SDP
Catalog No. 5000113553
Encompass_Preferred
The protein encoded by this gene is extracellular and enhances both neurite growth and neuronal survival. The encoded protein is found both as a GPI anchored membrane-bound form and as a secreted form. This activity-related ligand functions as a homodimer or heterodimer.
Catalog No. 50-001-13553 Supplier ABclonal Technology Supplier No. A165941000UL
The protein encoded by this gene is extracellular and enhances both neurite growth and neuronal survival. The encoded protein is found both as a GPI anchored membrane-bound form and as a secreted form. This activity-related ligand functions as a homodimer or heterodimer.