Learn More
ABclonal Technology CFAP53 Rabbit pAb
SDP
Catalog No. 5000109431
Encompass_Preferred
This gene belongs to the CFAP53 family. It was found to be differentially expressed by the ciliated cells of frog epidermis and in skin fibroblasts from human. Mutations in this gene are associated with visceral heterotaxy-6, which implicates this gene in determination of left-right asymmetric patterning.
Catalog No. 50-001-09431 Supplier ABclonal Technology Supplier No. A16607500UL
This gene belongs to the CFAP53 family. It was found to be differentially expressed by the ciliated cells of frog epidermis and in skin fibroblasts from human. Mutations in this gene are associated with visceral heterotaxy-6, which implicates this gene in determination of left-right asymmetric patterning.