Learn More
ABclonal Technology [KO Validated] GRK2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16621000UL
Encompass_Preferred
This gene encodes a member of the G protein-coupled receptor kinase family of proteins. The encoded protein phosphorylates the beta-adrenergic receptor as well as a wide range of other substrates including non-GPCR cell surface receptors, and cytoskeletal, mitochondrial, and transcription factor proteins. Data from rodent models supports a role for this gene in embryonic development, heart function and metabolism. Elevated expression of this gene has been observed in human patients with heart failure and Alzheimers disease.
Catalog No. 50-001-13556