Learn More
ABclonal Technology Cpsf4l Rabbit pAb
SDP
Catalog No. 5000111442
Encompass_Preferred
Predicted to be involved in pre-mRNA cleavage required for polyadenylation. Predicted to be part of mRNA cleavage and polyadenylation specificity factor complex. Is expressed in central nervous system and retina. Orthologous to human CPSF4L (cleavage and polyadenylation specific factor 4 like).
Catalog No. 50-001-11442 Supplier ABclonal Technology Supplier No. A16622500UL
Predicted to be involved in pre-mRNA cleavage required for polyadenylation. Predicted to be part of mRNA cleavage and polyadenylation specificity factor complex. Is expressed in central nervous system and retina. Orthologous to human CPSF4L (cleavage and polyadenylation specific factor 4 like).