Learn More
ABclonal Technology FCN2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A166901000UL
Encompass_Preferred
The product of this gene belongs to the ficolin family of proteins. This family is characterized by the presence of a leader peptide, a short N-terminal segment, followed by a collagen-like region, and a C-terminal fibrinogen-like domain. This gene is predominantly expressed in the liver, and has been shown to have carbohydrate binding and opsonic activities. Alternatively spliced transcript variants encoding different isoforms have been identified.
Catalog No. 50-001-11042