Learn More
ABclonal Technology MARK2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A167061000UL
Encompass_Preferred
This gene encodes a member of the Par-1 family of serine/threonine protein kinases. The protein is an important regulator of cell polarity in epithelial and neuronal cells, and also controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. The protein localizes to cell membranes. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-11555