Learn More
ABclonal Technology CDC25C Rabbit pAb
SDP
Supplier:  ABclonal Technology A1672500UL
Encompass_Preferred
This gene encodes a conserved protein that plays a key role in the regulation of cell division. The encoded protein directs dephosphorylation of cyclin B-bound CDC2 and triggers entry into mitosis. It also suppresses p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described.
Catalog No. 50-001-11325