Learn More
ABclonal Technology PEDF/SERPINF1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16731000UL
Encompass_Preferred
This gene encodes a member of the serpin family that does not display the serine protease inhibitory activity shown by many of the other serpin proteins. The encoded protein is secreted and strongly inhibits angiogenesis. In addition, this protein is a neurotrophic factor involved in neuronal differentiation in retinoblastoma cells. Mutations in this gene were found in individuals with osteogenesis imperfecta, type VI.
Catalog No. 50-001-11040