Learn More
ABclonal Technology SFRP5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A167341000UL
Encompass_Preferred
Secreted frizzled-related protein 5 (SFRP5) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP5 and SFRP1 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP5 is highly expressed in the retinal pigment epithelium, and moderately expressed in the pancreas.
Catalog No. 50-001-11021