Learn More
ABclonal Technology ADARB1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A167481000UL
Encompass_Preferred
This gene encodes the enzyme responsible for pre-mRNA editing of the glutamate receptor subunit B by site-specific deamination of adenosines. Studies in rat found that this enzyme acted on its own pre-mRNA molecules to convert an AA dinucleotide to an AI dinucleotide which resulted in a new splice site. Alternative splicing of this gene results in several transcript variants, some of which have been characterized by the presence or absence of an ALU cassette insert and a short or long C-terminal region.
Catalog No. 50-001-13366