Learn More
ABclonal Technology ARL4D Rabbit pAb
SDP
Supplier:  ABclonal Technology A167611000UL
Encompass_Preferred
ADP-ribosylation factor 4D is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4D is closely similar to ARL4A and ARL4C and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in membrane-associated intracellular trafficking.
Catalog No. 50-001-13359