Learn More
ABclonal Technology KAT5/Tip60 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16781000UL
Encompass_Preferred
The protein encoded by this gene belongs to the MYST family of histone acetyl transferases (HATs) and was originally isolated as an HIV-1 TAT-interactive protein. HATs play important roles in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and nonhistone proteins. This protein is a histone acetylase that has a role in DNA repair and apoptosis and is thought to play an important role in signal transduction. Alternative splicing of this gene results in multiple transcript variants.
Catalog No. 50-001-13399