Learn More
ABclonal Technology [KO Validated] DNA Polymerase beta Rabbit pAb
SDP
Supplier:  ABclonal Technology A1681500UL
Encompass_Preferred
The protein encoded by this gene is a DNA polymerase involved in base excision and repair, also called gap-filling DNA synthesis. The encoded protein, acting as a monomer, is normally found in the cytoplasm, but it translocates to the nucleus upon DNA damage. Several transcript variants of this gene exist, but the full-length nature of only one has been described to date.
Catalog No. 50-001-13401