Learn More
ABclonal Technology POU2F1/OCT1 Rabbit pAb
SDP
Catalog No. 5000113411
Encompass_Preferred
The OCT1 transcription factor was among the first identified members of the POU transcription factor family (summarized by Sturm et al., 1993 [PubMed 8314572]). Members of this family contain the POU domain, a 160-amino acid region necessary for DNA binding to the octameric sequence ATGCAAAT.
Catalog No. 50-001-13411 Supplier ABclonal Technology Supplier No. A1682500UL
The OCT1 transcription factor was among the first identified members of the POU transcription factor family (summarized by Sturm et al., 1993 [PubMed 8314572]). Members of this family contain the POU domain, a 160-amino acid region necessary for DNA binding to the octameric sequence ATGCAAAT.