Learn More
ABclonal Technology Pan Mono-Methyl lysine Rabbit pAb
SDP
Catalog No. 5000109479
Encompass_Preferred
Methylation of lysine residues is a common regulatory post-translational modification (PTM) that results in the mono-, di-, or tri-methylation of lysine at ε-amine groups by protein lysine methyltransferases (PKMTs).
Catalog No. 50-001-09479 Supplier ABclonal Technology Supplier No. A182931000UL
Methylation of lysine residues is a common regulatory post-translational modification (PTM) that results in the mono-, di-, or tri-methylation of lysine at ε-amine groups by protein lysine methyltransferases (PKMTs).