Learn More
Medchemexpress LLC A1874 | 50MG
SDP
Catalog No. 5000229321
Encompass_Preferred
A1874 is a nutlin-based (MDM2 ligand) and BRD4-degrading PROTAC with a DC50 of 32 nM (induce BRD4 degradation in cells) Effective in inhibiting many cancer cell lines proliferation[1]
Catalog No. 50-002-29321 Supplier Medchemexpress LLC Supplier No. HY11430550MG
A1874 is a nutlin-based (MDM2 ligand) and BRD4-degrading PROTAC with a DC50 of 32 nM (induce BRD4 degradation in cells) Effective in inhibiting many cancer cell lines proliferation[1]