Learn More
Enzo Life Sciences A1874 (5mg). CAS: 2064292-12-0
SDP
Catalog No. 502013729
Encompass_Preferred
A1874 is a nutlin-based PROTAC targeting on BRD4, consist of a BRD4-ligand JQ1 linked to a MDM2 E3 ligase ligand RG7388. A1874 possesses dual function as it dose-dependently degraded its target BRD4, accompanied with the downregulation of c-MYC, as well as stabilization of P53 and upregulated P21 due to the efficient engagement of MDM2 by RG7388. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-201-3729 Supplier Enzo Life Sciences Supplier No. ENZCHM2620005
A1874 is a nutlin-based PROTAC targeting on BRD4, consist of a BRD4-ligand JQ1 linked to a MDM2 E3 ligase ligand RG7388. A1874 possesses dual function as it dose-dependently degraded its target BRD4, accompanied with the downregulation of c-MYC, as well as stabilization of P53 and upregulated P21 due to the efficient engagement of MDM2 by RG7388. Purity: ≥98%. Long Term Storage: -20°C.