Learn More
BioVendor, LLC Alpha-1-Antitrypsin (stool, serum, plasma) Human ELISA (96 wells)
SDP
Catalog No. 504159
Encompass
Alpha-1-Antitrypsin. TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-HeparinSample Requirements50 µl/well (Diluted to 100X with buffer solution ©)ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below. Calibration Range2.5–160 ng/mlLimit of Detection0.198 ng/ml
Catalog No. 50-415-9 Supplier BioVendor, LLC Supplier No. IC6200
Alpha-1-Antitrypsin. TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-HeparinSample Requirements50 µl/well (Diluted to 100X with buffer solution ©)ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below. Calibration Range2.5–160 ng/mlLimit of Detection0.198 ng/ml