Learn More
Medchemexpress LLC A1AT modulator 1 | 2406205-61-4 | 99.8% | 370.42 | C21H23FN2O3 | 5 MG
SDP
Catalog No. 5000222048
Encompass_Preferred
A1AT modulator 1 is a research-grade small molecule inhibitor used to study Z α1-antitrypsin polymerization and related biochemical pathways. The compound is a solid (white to off-white) with molecular formula C21H23FN2O3 and molecular weight 370.42. It is provided for laboratory research use with storage recommendations for both powder and solution to maintain stability.
  • Potent inhibitor of Z α1-antitrypsin polymerization (pIC50 = 8.3).
  • High analytical purity (99.77%).
  • Solid, white to off-white appearance suitable for handling in lab settings.
  • Stable when stored under recommended conditions (powder: -20°C or 4°C; in solvent: -80°C or -20°C).
  • Provided in small-quantity packaging appropriate for research assays.
Catalog No. 50-002-22048 Supplier Medchemexpress LLC Supplier No. HY1348585MG
A1AT modulator 1 is a research-grade small molecule inhibitor used to study Z α1-antitrypsin polymerization and related biochemical pathways. The compound is a solid (white to off-white) with molecular formula C21H23FN2O3 and molecular weight 370.42. It is provided for laboratory research use with storage recommendations for both powder and solution to maintain stability.
  • Potent inhibitor of Z α1-antitrypsin polymerization (pIC50 = 8.3).
  • High analytical purity (99.77%).
  • Solid, white to off-white appearance suitable for handling in lab settings.
  • Stable when stored under recommended conditions (powder: -20°C or 4°C; in solvent: -80°C or -20°C).
  • Provided in small-quantity packaging appropriate for research assays.