Learn More
Aviva Systems Biology Corp A1CF Antibody - N-terminal region (ARP40778_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40778_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against A1CF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human A1CF. Alternate names: ACF, ACF64, ACF65, APOBEC1CF, ASP, MGC163391, RP11-564C4.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57622