Learn More
QA Bio Inc Ludger A1F Glycan - 20µg
Supplier:  QA Bio Inc CNA1F20U
Encompass
A1F Glycan Synonyms: A1F N-linked oligosaccharide. α(2,6)/FA2G2S(6)1, G2FS1, FA2G2S1, F(6)A2G2S1, F(6)A2G(4)2S(6)1 • Description:Mono-sialylated, core-fucosylated bi-antennary complex-type N-glycan (oligosaccharide). • Molecular Weight: 2079 • Purity: > 90% pure as assessed by a combination of 1 H-NMR and HPLC. • Sources: A1F glycan is found on many mammalian glycoproteins including IgG, gamma globulins, and many serum glycoproteins. This product is typically purified from the oligosaccharide pool released from porcine thyroglobulin by hydrazinolysis using a combination of HPLC and glycosidase digestion. • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. Contains ammonium salt to stabilise against desialylation.
Catalog No. 50-223-6586