Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich A1V2Pf1-Nhet-C10-Nh2

Catalog No. 91720650MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-720-650MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-720-650MG Supplier Sigma Aldrich Supplier No. 91720650MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protein degrader building block A1V2PF1-NHEt-C10-NH2 enables the synthesis of molecules for targeted protein degradation and SNIPER (specific and non-genetic inhibitor of apoptosis protein (IAP)-dependent protein erasers) technology. Developed in partnership with ComInnex, this conjugate contains an in silico-derived IAP-recruiting ligand, an alkyl-chain crosslinker, and a pendant amine for reactivity with an acid on a target warhead. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and protein degrader, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a terminal amine, including CRBN and VHL targeted, parallel synthesis can be used to more quickly generate SNIPER and PROTAC™ degrader libraries that feature variation in crosslinker length, composition, and E3 ligase ligand. Learn more about the novel IAP ligands generated through virtual screening of AVP mimetics in our Technology Spotlight. Building blocks in this series:917222 A1V2PF1-NHEt916943 A1V2PF1-NHEt-C6-NH2917206 A1V2PF1-NHEt-C10-NH2917451 A1V2PF1-NHEt-PEG1-NH2917702 A1V2PF1-NHEt-PEG3-NH2Technology Spotlight: Degrader Building Blocks with Inhibitor of Apoptosis Protein (IAP) In Silico-Derived LigandsIn Vivo Knockdown of Pathogenic Proteins via Specific and Nongenetic Inhibitor of Apoptosis Protein (IAP)-dependent Protein Erasers (SNIPERs)SNIPERs-Hijacking IAP activity to induce protein degradationE3 Ligase Ligands for PROTACs: How They Were Found and How to Discover New OnesPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

Molecular Formula C23H34N4O4S3
Linear Formula C23H34N4O4S3
Quantity 50 mg
Synonym (S)-1-((2S,5S)-17-Amino-2-cyclohexyl-5-methyl-4-oxo-9,12,15-trioxa-3,6-diazaheptadecanoyl)-N-((S)-1-(ethylamino)-1-oxo-3-phenylpropan-2-yl)pyrrolidine-2-carboxamide; AVP conjugate for IAP-mediated protein degrader development; SNIPER building block
Molecular Weight (g/mol) 674.87 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.