Learn More
Description
Specifications
Specifications
| CAS | 390040-83-0 |
| Linear Formula | C20H25N3O2 |
| Quantity | 50 mg |
| Synonym | tert-Butyl (17-(4-(4-(N-Benzyl-2-chloroacetamido)phenoxy)phenoxy)-3,6,9,12,15-pentaoxaheptadecyl)carbamate; Crosslinker-E3 Ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; RNF4-targeting building block; Template for synthesis of targeted protein degrader |
| Molecular Weight (g/mol) | 731.27 g/mol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.