Learn More
New England Biolabs, Inc. α2-3,6,8,9 Neuraminidase A - 4000 units
SDP
Catalog No. NC0824070
Encompass
a2-3,6,8,9 Neuraminidase A is a broad specificity sialidase, which cleaves linear and branched non-reducing terminal sialic acid residues from glycoproteins, glycopeptides, and oligosaccharides. It can be used for glycan analysis and characterization and intact glycoproteins.
Catalog No. NC0824070 Supplier New England Biolabs, Inc. Supplier No. P0722L
a2-3,6,8,9 Neuraminidase A is a broad specificity sialidase, which cleaves linear and branched non-reducing terminal sialic acid residues from glycoproteins, glycopeptides, and oligosaccharides. It can be used for glycan analysis and characterization and intact glycoproteins.