Learn More
Athens Research And Technology Inc Alpha 2 Antiplasmin (SERPINF2), Native Human Plasma, ≥95% (SDS-PAGE), 1 mg
Catalog No. 50489610
Encompass_Preferred
Alpha 2 Antiplasmin (SERPINF2) is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied lyophilized in a 1 mg pack. Molecular weight: 70 kDa. Alpha 2 Antiplasmin, Human Plasma: 1 MG; Lyophilized from 20 mM Bis-Tris, pH 6.4, with 200 mM NaCl; MW 70,000; EC= 0.7; Activity: > = 5.0 IU/mg; Purity: > =95% by SDS-PAGE; Inhibitor, Coagulation Factor.
Catalog No. 50-489-610 Supplier Athens Research And Technology Inc Supplier No. 16160129011MG
Alpha 2 Antiplasmin (SERPINF2) is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied lyophilized in a 1 mg pack. Molecular weight: 70 kDa. Alpha 2 Antiplasmin, Human Plasma: 1 MG; Lyophilized from 20 mM Bis-Tris, pH 6.4, with 200 mM NaCl; MW 70,000; EC= 0.7; Activity: > = 5.0 IU/mg; Purity: > =95% by SDS-PAGE; Inhibitor, Coagulation Factor.