Learn More
Vici Precision Sampling, Inc. A2 C & D NEEDLES PK/3
Catalog No. NC9373416
Encompass_Preferred
Needles Gas A2, C & D, .028 in. x .005 in. x 2 in. (22 gauge) bevel open end
Catalog No. NC9373416 Supplier Vici Precision Sampling, Inc. Supplier No. 943050
Needles Gas A2, C & D, .028 in. x .005 in. x 2 in. (22 gauge) bevel open end