Learn More
Vici Precision Sampling, Inc. A-2 Series Syringes, 50 uL, Standard
Catalog No. NC1634744
Encompass_Preferred
.029" x .005" x 2" Needle size. The A-2 features a push-button valve, which allows sample storage to 250 psi in syringes as small as 25 µl. This is useful with small liquid samples containing low-boiling components which would be lost through evaporation using ordinary syringes.
Catalog No. NC1634744 Supplier Vici Precision Sampling, Inc. Supplier No. 050024
.029" x .005" x 2" Needle size. The A-2 features a push-button valve, which allows sample storage to 250 psi in syringes as small as 25 µl. This is useful with small liquid samples containing low-boiling components which would be lost through evaporation using ordinary syringes.