Learn More
Athens Research And Technology Inc A2 MACROGLOBULIN HU 1MG
Supplier:  Athens Research And Technology Inc 16160120131MG
Encompass_Preferred
Alpha 2 Macroglobulin, Human Plasma, 1 MG: Lyophilized from 30 mM Na Phosphate, pH 7.0 & 1 mg glycine per 1 mg Alpha 2 Macroglobulin; MW 725,000; EC=0.81; Purity: > =95% by SDS-PAGE; Native; Purified from human plasma in Athens Research & Tech lab.
Catalog No. 50-489-615