Learn More
Enzo Life Sciences A20 monoclonal antibody (59A426) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM009E
Encompass_Preferred
Clone: 59A426. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human A20. Species reactivity: Human. Applications: IHC (PS), IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-7130