Learn More
Cayman Chemical A20FMDV2trIfluoROCTAte sa 10mg
SDP
Catalog No. 502915447
Encompass_Preferred
A peptide; inhibits αVβ6 integrin-dependent cell adhesion in H357 cells expressing human αVβ6 (IC50 = 1.2 µM); When coupled with 111indium-DTPA, A20FMDV2 binds to αVβ6 integrin endogenously expressed for radiation imaging analysis in a MCF10CA1a breast cancer mouse xenograft model at 20 MBq
Catalog No. 50-291-5447 Supplier Cayman Chemical Supplier No. 3695810 MG
A peptide; inhibits αVβ6 integrin-dependent cell adhesion in H357 cells expressing human αVβ6 (IC50 = 1.2 µM); When coupled with 111indium-DTPA, A20FMDV2 binds to αVβ6 integrin endogenously expressed for radiation imaging analysis in a MCF10CA1a breast cancer mouse xenograft model at 20 MBq