Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGuard™ A22A1 Series Powdered Vinyl 3.0 mil Gloves, Clear

Catalog No. 17004392
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-392 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-004-392 Supplier VGuard™ Supplier No. A22A11
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

A22 Clear Vinyl Industrial Gloves resist abrasions, cuts and punctures while offering a comfortable fit and feel.

  • Vinyl
  • Powdered
  • Non-Sterile
  • Ambidextrous
  • Color: Clear
  • Texture: Smooth
  • Palm Thickness: 2.8 mil
  • Fingertip Thickness: 3.0 mil
  • Length: Standard (23 cm)
  • Cuff: Beaded
  • FDA Compliant for foodservice applications.
  • 100 Gloves/Box

Specifications

Accelerator Free No
Applications Industrial
AQL 1.5
Color Clear
Disposable Single-use
Exterior Finish No Texture
Finger Thickness (English) 3.0 mil
Latex Free Yes
Material Vinyl
Palm Thickness (English) 2.8 mil
Powdered Powdered
Size Small
Sterility Non-sterile
Thickness (English) 3.0 mil
Glove Cuff Style Beaded
Model A22A11
Series Vanguard Safety
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.