Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A23187, 98%, free acid, Thermo Scientific Chemicals

Catalog No. AC328080050
Change view
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Glass Bottle
Glass Bottle in Plastic Container
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC328080050 5 mg Glass Bottle
AC328080010 1 mg Glass Bottle in Plastic Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC328080050 Supplier Thermo Scientific Chemicals Supplier No. 328080050
  • $267.00
    $216.65 / Each of 1

    Save $50.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

A23187 (Calcimycin), CAS # 52665-69-7, is a calcium ionophore-based antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description


• A23187 is a polyether substance that permeabilizes cell membranes to calcium and other divalent cations


Applications
• A23187 can be used for the study of mitochondrial physiology because it acts as an uncoupling agent
• A23187 can be used to study the roles of divalent cations in cell physiology
• When bound to fluorescent calcium ions, A23187 can be useful for imaging studies

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52665-69-7
Molecular Formula C29H37N3O6
Molecular Weight (g/mol) 523.63
MDL Number MFCD00151202,MFCD00151202
InChI Key HIYAVKIYRIFSCZ-CVXKHCKVSA-N
PubChem CID 97043288
IUPAC Name 5-(methylamino)-2-[[(2R,3S,5S,6S,8S,9S)-3,5,9-trimethyl-2-[(2S)-1-oxo-1-(1H-pyrrol-2-yl)propan-2-yl]-1,7-dioxaspiro[5.5]undecan-8-yl]methyl]-1,3-benzoxazole-4-carboxylic acid
SMILES CNC1=CC=C2OC(C[C@@H]3OC4(CC[C@@H]3C)O[C@H](C(C)C(=O)C3=CC=CN3)[C@H](C)C[C@H]4C)=NC2=C1C(O)=O

Specifications

Physical Form Solid
CAS Min % 97.5
Melting Point 182.0°C to 193.0°C
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass Bottle
Color White
Quantity 5 mg
Assay Percent Range 98%
Merck Index 15, 1642
Solubility Information Solubility in water: .. Other solubilities: soluble in dmso, ethylacetate, methanol, ethanol
Formula Weight 523.63
Percent Purity 98%
Chemical Name or Material A23187, Free acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.